Merck
  • Ex vivo microbial leakage after using different final irrigation regimens with chlorhexidine.
Ex vivo microbial leakage after using different final irrigation regimens with chlorhexidine.
Journal of applied oral science : revista FOB (2013-04-06)
Esther Navarro-Escobar, Pilar Baca, María Paloma González-Rodríguez, María Teresa Arias-Moliz, Matilde Ruiz, Carmen María Ferrer-Luque
ABSTRACT
To assess the influence of final irrigation protocols with chlorhexidine in the coronal leakage of Enterococcus faecalis in filled root canals. Seventy single-root canals from extracted teeth were prepared using ProTaper instruments. The irrigation protocol accomplished an alternating irrigation with 5 mL of 2.5% sodium hypochlorite (NaOCI) and 17% EDTA between each file. The teeth were randomly divided into four experimental groups (n=15) according to the final irrigation regimen: group 1, without final irrigation; group 2, irrigation with 10 mL 2.0% chlorhexidine (CHX); group 3, with a final application of EC40™; and group 4, irrigation with the combination (1:1) of 0.2% CHX + 0.1% cetrimide (CTR). All the teeth were mounted in a two-chamber apparatus and the coronal access was exposed to E. faecalis. The presence of turbidity in the BHI broth over a period of 180 days was observed. The Friedman test was used for statistical analysis. EC40™ varnish showed the least leakage at 180 days, and was statistically similar to 2% CHX. No significant differences were observed between the group without final irrigation and the 2% CHX group or 0.2% CHX + 0.1% CTR. In this ex vivo study, EC40™ showed the longest delayed coronal leakage of E. faecalis, although without significant differences from 2% CHX.
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Cetyltrimethylammonium hydrogensulfate, 99%
Chlorhexidine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Supelco
Hexadecyltrimethylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Chlorhexidine, ≥99.5%
Sigma-Aldrich
Chlorhexidine dihydrochloride, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Chlorhexidine, European Pharmacopoeia (EP) Reference Standard
Supelco
Hexadecyltrimethylammonium bromide, analytical standard