Merck
  • IL-33-mediated innate response and adaptive immune cells contribute to maximum responses of protease allergen-induced allergic airway inflammation.
IL-33-mediated innate response and adaptive immune cells contribute to maximum responses of protease allergen-induced allergic airway inflammation.
Journal of immunology (Baltimore, Md. : 1950) (2013-04-03)
Seiji Kamijo, Haruna Takeda, Tomoko Tokura, Mayu Suzuki, Kyoko Inui, Mutsuko Hara, Hironori Matsuda, Akira Matsuda, Keisuke Oboki, Tatsukuni Ohno, Hirohisa Saito, Susumu Nakae, Katsuko Sudo, Hajime Suto, Saori Ichikawa, Hideoki Ogawa, Ko Okumura, Toshiro Takai
ABSTRACT
How the innate and adaptive immune systems cooperate in the natural history of allergic diseases has been largely unknown. Plant-derived allergen, papain, and mite allergens, Der f 1 and Der p 1, belong to the same family of cysteine proteases. We examined the role of protease allergens in the induction of Ab production and airway inflammation after repeated intranasal administration without adjuvants and that in basophil/mast cell stimulation in vitro. Papain induced papain-specific IgE/IgG1 and lung eosinophilia. Der f 1 induced Der f 1-specific IgG1 and eosinophilia. Although papain-, Der f 1-, and Der p 1-stimulated basophils expressed allergy-inducing cytokines, including IL-4 in vitro, basophil-depleting Ab and mast cell deficiency did not suppress the papain-induced in vivo responses. Protease inhibitor-treated allergens and a catalytic site mutant did not induce the responses. These results indicate that protease activity is essential to Ab production and eosinophilia in vivo and basophil activation in vitro. IL-33-deficient mice lacked eosinophilia and had reduced papain-specific IgE/IgG1. Coadministration of OVA with papain induced OVA-specific IgE/IgG1, which was reduced in IL-33-deficient mice. We demonstrated IL-33 release, subsequent IL-33-dependent IL-5/IL-13 release, and activation of T1/ST2-expressing lineage(-)CD25(+)CD44(+) innate lymphoid cells in the lung after papain inhalation, suggesting the contribution of the IL-33-type 2 innate lymphoid cell-IL-5/IL-13 axis to the papain-induced airway eosinophilia. Rag2-deficient mice, which lack adaptive immune cells, showed significant, but less severe, eosinophilia. Collectively, these results suggest cooperation of adaptive immune cells and IL-33-responsive innate cells in protease-dependent allergic airway inflammation.
Sigma-Aldrich
Papain from papaya latex, crude powder, 1.5-10 units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Papain from papaya latex, lyophilized powder, aseptically filled
Pricing and availability is not currently available.
Sigma-Aldrich
Papain from Carica papaya, solution, light brown, ≥10 U/mg protein (~25 mg/ml)
Pricing and availability is not currently available.