Merck
Lactic acid oligomers (OLAs) as prodrug moieties.
Die Pharmazie (2013-03-09)
J Kruse, B Lachmann, R Lauer, S Eppacher, C R Noe
ABSTRACT
In this paper we propose the use of lactic acid oligomers (OLAs) as prodrug moieties. Two synthetic approaches are presented, on the one hand a non selective oligomerisation of lactic acid and on the other hand a block synthesis to tetramers of lactic acid. Dimers of lactic acid were investigated with respect to their plasma stability and their adsorption to albumine. Ibuprofen was chosen as the first drug for OLAylation. The ester 19 of LA(1)-ibuprofen was evaluated with respect to the degradation to human plasma and the adsorption to albumine. All results indicate that lactic acid oligomers are promising prodrug moieties.
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%
Supelco
Ibuprofen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ibuprofen sodium salt, ≥98% (GC)
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard