Merck
  • A synthetic dolastatin 10 analogue suppresses microtubule dynamics, inhibits cell proliferation, and induces apoptotic cell death.
A synthetic dolastatin 10 analogue suppresses microtubule dynamics, inhibits cell proliferation, and induces apoptotic cell death.
Journal of medicinal chemistry (2013-03-01)
Praveen Kumar Gajula, Jayant Asthana, Dulal Panda, Tushar Kanti Chakraborty
ABSTRACT
We have synthesized eight analogues (D1-D8) of dolastatin 10 containing several unique amino acid subunits. Of these agents, D5 was found to be most effective in inhibiting both HeLa cell proliferation and microtubule assembly in vitro. At low nanomolar concentrations, D5 inhibited the proliferation of several types of cancer cells in culture. D5 bound to tubulin with a dissociation constant of 29.4 ± 6 μM. D5 depolymerized microtubules in cultured cells and produced mulitpolar spindles. At its half-maximal inhibitory concentration (15 nM), D5 strongly suppressed the dynamics of individual microtubules in live MCF-7 cells. D5 increased the accumulation of checkpoint proteins BubR1 and Mad2 at the kinetochoric region and caused G2/M block in these cells. The blocked cells underwent apoptosis with the activation of Jun N-terminal kinase. The results suggested that D5 exerts its antiproliferative action by dampening microtubule dynamics.
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
Pricing and availability is not currently available.