Merck
  • Nanostructured manganese oxide particles from coordination complex decomposition and their catalytic properties for ethanol oxidation.
Nanostructured manganese oxide particles from coordination complex decomposition and their catalytic properties for ethanol oxidation.
Journal of nanoscience and nanotechnology (2013-02-21)
Humberto Palza, Andrés Maturana, Francisco Gracia, Andrónico Neira, Victor M Fuenzalida, Jonathan Avila, Noelia M Sanchez-Ballester, Mark R J Elsegood, Simon J Teat, Katsuhiko Ariga, Jonathan P Hill
ABSTRACT
Novel manganese oxide particles with complex morphologies and different nanostructures (i.e., spherical/lamellar) were synthesized by initial preparation of a coordination complex of manganese with 1,4,7,10-tetraazacyclododecane (cyclen), followed by characterization of the nanostructured oxide as a catalytic material for ethanol oxidation. The samples present a bulk gamma-MnO2 structure although X-ray photoelectron spectroscopy analysis reveals that their surfaces have different chemical compositions. Some of these nanostructured particles show high catalytic activities for ethanol oxidation enabling a decrease of the reaction temperature by more than 80 degrees C as compared with traditional MnO2 particles. The high catalytic activity of the particles depends on their morphology and a relationship between morphology and specific area was established. It is proposed that these novel nanostructured manganese oxide particles may be highly active in the catalytic oxidation of other volatile organic compounds (VOCs) opening up their further development for environmental applications.
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh
Sigma-Aldrich
Magnesium oxide, light, 95%