Merck
  • Emergency euthanasia of cattle challenged with Escherichia coli O157:H7 - a case study for evaluating the response to an infectious disease outbreak.
Emergency euthanasia of cattle challenged with Escherichia coli O157:H7 - a case study for evaluating the response to an infectious disease outbreak.
Journal of veterinary science (2013-02-08)
Brandon H Gilroyed, Tim Reuter, John P Kastelic, Tim A McAllister
ABSTRACT
In the event of an infectious disease outbreak in cattle, carcasses must be disposed of in a rapid and contained manner. This brief communication details injection of a barbiturate to euthanize cattle inoculated with Escherichia coli O157:H7 followed by carcass composting in a manner that prevents the spread of infectious agents.
Product Description
Supelco
Pentobarbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®