Skip to Content
End of year dates: Orders must be billed by 05/12. Merck Mexico will be closed from 18/12 to 02/01.
Merck
Pharmacologic treatment of pediatric headaches: a meta-analysis.
JAMA pediatrics (2013-01-30)
Khalil El-Chammas, Jill Keyes, Nathan Thompson, Jayanthi Vijayakumar, Dorothy Becher, Jeffrey L Jackson
ABSTRACT
OBJECTIVE To assess the effectiveness of prophylactic headache treatment in children and adolescents. DATA SOURCES PubMed, EMBASE, Cochrane Database of Clinical Trials, and bibliography of retrieved articles through August 11, 2012. STUDY SELECTION Randomized trials of headache treatment among children and adolescents (<18 years old). INTERVENTION Any placebo-controlled trial or comparisons between 2 or more active medications. MAIN OUTCOME MEASURE Number of headaches per month. RESULTS Among 21 included trials, there were 13 placebo-controlled and 10 active comparator trials (2 also included placebo). Twenty trials focused on episodic migraines and 1 on chronic daily headaches. Drugs more effective than placebo for episodic migraines (<15 headaches per month) included topiramate (difference in headaches per month, -0.71; 95% CI, -1.19 to -0.24) and trazodone (-0.60; 95% CI, -1.09 to -0.11). Ineffective drugs included clonidine, flunarizine, pizotifen, propranolol, and valproate. A single trial of fluoxetine for chronic daily headaches found it ineffective. Patients given placebo experienced a significant (P = .03) decline in headaches, from 5.6 (95% CI, 4.52-6.77; Q = 8.14 [Cochran Q is a measure of the heterogeneity of the included studies]) to 2.9 headaches per month (95% CI, 1.66-4.08; Q = 4.72). Among the 10 active comparator trials, flunarizine was more effective than piracetam (difference in headaches per month, -2.20; 95% CI, -3.93 to -0.47) but no better than aspirin, dihydroergotamine, or propranolol. Propranolol was compared with valproate as well as behavioral treatment, and 2 studies compared different doses of topiramate; none of these trials showed significant differences. CONCLUSIONS Topiramate and trazodone have limited evidence supporting efficacy for episodic migraines. Placebo was effective in reducing headaches. Other commonly used drugs have no evidence supporting their use in children and adolescents. More research is needed.
Product Description
Supelco
Trazodone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Sigma-Aldrich
Trazodone hydrochloride, ≥99% (HPLC), powder
Pricing and availability is not currently available.