Merck
  • Enantioselective formal aza-Diels-Alder reactions of enones with cyclic imines catalyzed by primary aminothioureas.
Enantioselective formal aza-Diels-Alder reactions of enones with cyclic imines catalyzed by primary aminothioureas.
Journal of the American Chemical Society (2013-01-17)
Mathieu P Lalonde, Meredeth A McGowan, Naomi S Rajapaksa, Eric N Jacobsen
ABSTRACT
A highly enantio- and diastereoselective synthesis of indolo- and benzoquinolizidine compounds has been developed through the formal aza-Diels-Alder reaction of enones with cyclic imines. This transformation is catalyzed by a new bifunctional primary aminothiourea that achieves simultaneous activation of both the enone and imine reaction components.
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Pricing and availability is not currently available.