Merck
  • Design of a new fluorescent probe: pyrrole/imidazole hairpin polyamides with pyrene conjugation at their γ-turn.
Design of a new fluorescent probe: pyrrole/imidazole hairpin polyamides with pyrene conjugation at their γ-turn.
Bioorganic & medicinal chemistry (2013-01-15)
Thangavel Vaijayanthi, Toshikazu Bando, Kaori Hashiya, Ganesh N Pandian, Hiroshi Sugiyama
ABSTRACT
Fluorophores that are conjugated with N-methylpyrrole-N-methylimidazole (Py-Im) polyamides postulates versatile applications in biological and physicochemical studies. Here, we show the design and synthesis of new types of pyrene-conjugated hairpin Py-Im polyamides (1-5). We evaluated the steady state fluorescence of the synthesized conjugates (1-5) in the presence and absence of oligodeoxynucleotides 5'-CGTATGGACTCGG-3' (ODN 1) and 5'-CCGAGTCCATACG-3' (ODN 2) and observed a distinct increase in emission at 386nm with conjugates 4 and 5. Notably, conjugate 5 that contains a β-alanine linker had a stronger binding affinity (K(D)=1.73×10(-8)M) than that of conjugate 4 (K(D)=1.74×10(-6)M). Our data suggests that Py-Im polyamides containing pyrene fluorophore with a β-alanine linker at the γ-turn NH(2) position can be developed as the competent fluorescent DNA-binding probes.
Product Description
Supelco
Pyrene, analytical standard
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
Imidazole hydrochloride
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole sodium derivative, technical grade
Imidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O