Merck
  • The effect of phytic acid on the expression of NF-kappaB, IL-6 and IL-8 in IL-1beta-stimulated human colonic epithelial cells.
The effect of phytic acid on the expression of NF-kappaB, IL-6 and IL-8 in IL-1beta-stimulated human colonic epithelial cells.
Acta poloniae pharmaceutica (2013-01-05)
Joanna Wawszczyk, Małgorzata Kapral, Andrzej Hollek, Ludmiła Weglarz
ABSTRACT
Intestinal epithelial cells play an important role in the mucosal immune and inflammatory reactions via the expression and secretion of proinflammatory cytokines such as interleukin-6 (IL-6) and interleukin-8 (IL-8). The expression of both interleukins is regulated by nuclear factor KB (NF-kappaB). Phytic acid (IP6) is an essential component of high fiber diet. It is physiologically present in the human large gut at concentrations reaching 4 mM. It exhibits pleiotropic health beneficial effects including anti-oxidant and anti-tumor activities. Recent studies showed that IP6 can modulate immune functions of intestinal epithelium through regulation of proinflammatory cytokines secretion. The aim of this study was to analyze the effect of IP6 on the expression of IL-6 and IL-8 as well as p50 and p65 subunits of NF-kappaB and its inhibitor IkappaBalpha in Caco-2 cells stimulated with IL-1beta. A kinetic study of mRNAs expression in cells was performed after their treatment with 1 and 2.5 mM IP6 for 3, 6 and 12 h. Quantification of the genes expression was carried out using real time QRT-PCR technique. IP6 at all used concentrations had no influence on transcription of p65 gene and modulated expression of p50 and IkappaBalpha genes in Caco-2 cells. Treatment of cells with IP6 resulted in a marked decrease in both IL-6 (at 3 and 6 h) and IL-8 expression (3 h). The results of these studies suggest that IP6 may exert immunoregulatory effects on intestinal epithelium by influencing transcriptional activity of genes encoding p50 subunit of NF-kappaB, its inhibitor IkappaBalpha and proinflammatory cytokines IL-6 and IL-8.
Sigma-Aldrich
Phytic acid solution, 50 % (w/w) in H2O