Merck
  • Chemoselective nitration of aromatic sulfonamides with tert-butyl nitrite.
Chemoselective nitration of aromatic sulfonamides with tert-butyl nitrite.
Chemical communications (Cambridge, England) (2012-12-01)
Brenden Kilpatrick, Markus Heller, Steve Arns
ABSTRACT
A methodology for the efficient conversion of aromatic sulfonamides into their mono-nitro derivatives using tert-butyl nitrite is reported. The reaction exhibits a high degree of chemoselectivity for sulfonamide functionalized aryl systems, even in the presence of other sensitive or potentially reactive functionalities.
Sigma-Aldrich
Butyl nitrite, 95%
Sigma-Aldrich
tert-Butyl nitrite, technical, ≥90% (GC)