Merck
  • Investigation and enhancement of the stability and performance of water reduction systems based on cyclometalated iridium(III) complexes.
Investigation and enhancement of the stability and performance of water reduction systems based on cyclometalated iridium(III) complexes.
ChemSusChem (2012-11-14)
Sven Hansen, Marga-Martina Pohl, Marcus Klahn, Anke Spannenberg, Torsten Beweries
ABSTRACT
Water reduction systems that use a bis-cyclometalated Ir(III) photosensitiser (PS) along with homogeneous Pd complexes as a source of in-situ-formed colloidal Pd as the water reducing complex (WRC) and triethylamine (TEA) as the sacrificial electron donor were tested and characterised with respect to their photocatalytic H(2) production performance. It was confirmed that substitution of the 2-(pyridin-2-yl)benzen-1-ide (pyb) ligand in the well-known system [Ir(pyb)(2)(bpy)](+) (bpy=2,2'-bipyridine) by the fluorinated cyclometalating ligand 5-fluoro-2-(5-methylpyridin-2-yl)benzen-1-ide (Fmpyb) tremendously enhanced the H(2) production rate. Moreover, variation of the bidentate N^N ligand bpy by alkyl substitution in the 4,4'-position resulted in an increase in the H(2) production yield by a factor of three. The incident-photon-to-hydrogen-efficiency could be enhanced from 2.6 to 12.3%. Furthermore, a new dinuclear Co complex was used as a reduction catalyst and showed up to 760 turnovers after 20 h. A detailed study of the concentration impact of all components in the photoredox system was performed. DFT calculations were used to aid the explanation of the findings.
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine hydrochloride, ≥99.0% (AT)