Merck
  • Distribution variation of a metabolic uncoupler, 2,6-dichlorophenol (2,6-DCP) in long-term sludge culture and their effects on sludge reduction and biological inhibition.
Distribution variation of a metabolic uncoupler, 2,6-dichlorophenol (2,6-DCP) in long-term sludge culture and their effects on sludge reduction and biological inhibition.
Water research (2012-11-06)
Yu Tian, Jun Zhang, Di Wu, Zhipeng Li, Yanni Cui
ABSTRACT
Distribution variation of a metabolic uncoupler, 2,6-dichlorophenol (2,6-DCP), in long-term sludge culture was studied, and the effects on sludge reduction and biological inhibition of this chemical during the 90-day operation were established. The extracellular polymeric substance (EPS) matrix functioned as a protective barrier for the bacteria inside sludge flocs to 2,6-DCP, resulting in the transfer of 2,6-DCP from the liquid phase to the activated sludge fraction. Significant sludge reduction (about 40%) was observed after the addition of 2,6-DCP in the first 40 days, while the ineffective function of 2,6-DCP in sludge reduction (days 70-90) might be correlated to the EPS protection mechanism. The inhibitory effect of 2,6-DCP on the COD removal was extremely lower than on the nitrification performance due to the fact that 2,6-DCP was much more toxic to autotrophic microorganisms than heterotrophic microorganisms. Moreover, both of them recovered to a higher level again with the transfer potential of 2,6-DCP to sludge. Thus, the application of metabolic uncoupler for excess sludge reduction should be cautious.
Sigma-Aldrich
2,6-Dichlorophenol, 99%