Merck
  • Valence band-edge engineering of nickel oxide nanoparticles via cobalt doping for application in p-type dye-sensitized solar cells.
Valence band-edge engineering of nickel oxide nanoparticles via cobalt doping for application in p-type dye-sensitized solar cells.
ACS applied materials & interfaces (2012-10-12)
Gayatri Natu, Panitat Hasin, Zhongjie Huang, Zhiqiang Ji, Mingfu He, Yiying Wu
ABSTRACT
We have systematically studied the effects of substitutional doping of p-type nanoparticulate NiO with cobalt ions. Thin films of pure and Co-doped NiO nanoparticles with nominal compositions Co(x)Ni(1-x)O(y) (0 ≤ x ≤ 0.1) were fabricated using sol-gel method. X-ray photoelectron spectroscopy revealed a surface enrichment of divalent cobalt ions in the Co(x)Ni(1-x)O(y) nanoparticles. Mott-Schottky analysis in aqueous solutions was used to determine the space charge capacitance values of the films against aqueous electrolytes, which yielded acceptor state densities (N(A)) and apparent flat-band potentials (E(fb)). Both N(A) and E(fb) values of the doped NiO were found to gradually increase with increasing amount of doping; thus the Fermi energy level of the charge carriers decreased with Co-doping. The photovoltage of p-DSCs constructed using the Co(x)Ni(1-x)O(y) films increased with increasing amount of cobalt, as expected from the trend in the E(fb). Co-doping increased both carrier lifetimes within the p-DSCs and the carrier transport times within the nanoparticulate semiconductor network. The nominal composition of Co₀.₀₆Ni₀.₉₄O(y) was found to be optimal for use in p-DSCs.
Sigma-Aldrich
Nickel(II) oxide, nanopowder, <50 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Nickel(II) oxide, ≥99.995% trace metals basis
Sigma-Aldrich
Nickel(II) oxide, 99.99% trace metals basis
Sigma-Aldrich
Nickel(II) oxide, green, −325 mesh, 99%