Merck
  • Synthesis of phenol derivatives from cyclohex-2-enones bearing an alkyne through Lewis acid-catalyzed enolization and intramolecular Alder-Rickert reaction.
Synthesis of phenol derivatives from cyclohex-2-enones bearing an alkyne through Lewis acid-catalyzed enolization and intramolecular Alder-Rickert reaction.
The Journal of organic chemistry (2012-09-26)
Atsushi Kinbara, Takehiro Yamagishi, Naoko Hanzawa, Etsuko Kawashima, Hiroaki Miyaoka
ABSTRACT
A cationic rhodium(I) complex- or In(OTf)(3)-catalyzed synthesis of phenol derivatives from cyclohex-2-enone having an ethoxycarbonyl-substituted alkyne has been achieved. This reaction proceeds via enolization and an intramolecular Alder-Rickert reaction.
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis