Merck
  • Synthesis of bicyclic imidazoles via [2 + 3] cycloaddition between nitriles and regioselectively generated α-imino gold carbene intermediates.
Synthesis of bicyclic imidazoles via [2 + 3] cycloaddition between nitriles and regioselectively generated α-imino gold carbene intermediates.
Organic letters (2012-08-25)
Yuanjing Xiao, Liming Zhang
ABSTRACT
The cyclic α-imino gold carbene intermediate B is most likely generated in situ via regioselective nitrene transfer from an azido group to a tethered terminal alkyne in the presence of a gold catalyst and at ambient temperature. This highly electrophilic intermediate can react with a weakly nucleophilic nitrile, which is used as the reaction solvent, to deliver a bicyclic imidazole rapidly in an overall bimolecular [2 + 2 + 1] cycloaddition and in mostly serviceable yield. The competing intramolecular Huisgen reaction, although likely also catalyzed by gold, is minimized by using AuCl(3) as the catalyst.
Sigma-Aldrich
Gold(III) chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Gold(III) chloride, 99%
Sigma-Aldrich
Gold(I) chloride, 99.9% trace metals basis