Merck
  • Chemoselective nanoporous membranes via chemically directed assembly of nanoparticles and dendrimers.
Chemoselective nanoporous membranes via chemically directed assembly of nanoparticles and dendrimers.
Advanced materials (Deerfield Beach, Fla.) (2012-08-24)
Myoung-Hwan Park, Chandramouleeswaran Subramani, Subinoy Rana, Vincent M Rotello
ABSTRACT
Chemoselective nanoporous membranes: tunably porous membranes with embedded functionalities are generated using a template-free, chemically directed nanoparticle-dendrimer (NP-Den) network assembly. This approach provides a direction in the design of post-functionalizable nanoporous membranes for distinguishing both organic molecules and proteins with excellent chemo- and bioselectivity.
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Rhodamine 6G perchlorate, Dye content 99 %
Supelco
Rhodamine 6G, analytical standard