Merck
  • Effects of gemfibrozil on lipid metabolism, steroidogenesis, and reproduction in the fathead minnow (Pimephales promelas).
Effects of gemfibrozil on lipid metabolism, steroidogenesis, and reproduction in the fathead minnow (Pimephales promelas).
Environmental toxicology and chemistry (2012-08-23)
Sarah Y Skolness, Elizabeth J Durhan, Kathleen M Jensen, Michael D Kahl, Elizabeth A Makynen, Daniel L Villeneuve, Gerald T Ankley
ABSTRACT
Fibrates are a class of pharmaceuticals that indirectly modulate cholesterol biosynthesis through effects on peroxisome proliferator-activated receptors. Gemfibrozil is a fibrate that has been detected in wastewater treatment plant influents, effluents, and drinking water. The objective of the present study was to assess the potential physiological and reproductive impacts of gemfibrozil on fathead minnows (Pimephales promelas). Fish were exposed to gemfibrozil in two different studies. The first was a short-term test with water concentrations of 0, 15, and 600 µg gemfibrozil/L, sampling after 2 or 8 d of exposure. Plasma cholesterol concentrations were significantly reduced in males exposed to 600 µg gemfibrozil/L for 8 d. In addition, expression of several hepatic genes important to lipid metabolism was altered, suggesting that gemfibrozil does affect lipid metabolism in fish. A 21-d study was conducted to investigate further the effects on lipid metabolism and steroidogenesis as well as to assess potential impacts of gemfibrozil on reproduction. Fish were exposed to water concentrations of 0, 1.5, 15, 600, and 1,500 µg gemfibrozil/L. Exposure to 1,500 µg gemfibrozil/L caused a modest, but not significant, reduction in fecundity. However, gemfibrozil had no consistent effect on plasma cholesterol, triglycerides, or sex steroids after 21 d of exposure. The present study showed no evidence for significant physiological or reproductive impacts of gemfibrozil at an environmentally relevant concentration of 1.5 µg/L.
Product Description
Gemfibrozil, European Pharmacopoeia (EP) Reference Standard
Gemfibrozil for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Gemfibrozil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Gemfibrozil