Merck
  • Modified coaxial electrospinning for the preparation of high-quality ketoprofen-loaded cellulose acetate nanofibers.
Modified coaxial electrospinning for the preparation of high-quality ketoprofen-loaded cellulose acetate nanofibers.
Carbohydrate polymers (2012-07-31)
Deng-Guang Yu, Jia-Hui Yu, Lan Chen, Gareth R Williams, Xia Wang
ABSTRACT
This study investigates the use of a modified coaxial electrospinning process in the production of drug-loaded cellulose acetate (CA) nanofibers. With CA employed as a filament-forming matrix and ketoprofen (KET) as an active pharmaceutical ingredient, modified coaxial processes using sheath fluids comprising only mixed solvents were undertaken. With a sheath-to-core flow rate ratio of 0.2:1, the nanofibers prepared from the coaxial process had a smaller average diameter, narrower size distribution, more uniform structures, and smoother surface morphologies than those generated from single fluid electrospinning. In addition, the coaxial fibers provided a better zero-order drug release profile. The use of a sheath solvent means that the core jet is subjected to electrical drawing for a longer period, facilitating homogeneous core jet solidification and retarding the formation of wrinkles on the surface of the nanofibers. This modified coaxial electrospinning protocol allows the systematic fabrication of functional polymer nanofibers with improved quality.
Sigma-Aldrich
Cellulose acetate, average Mn ~50,000 by GPC
Sigma-Aldrich
Cellulose acetate, average Mn ~30,000 by GPC