Merck
A photochemical route to 2-substituted benzo[b]furans.
The Journal of organic chemistry (2012-07-11)
Stefano Protti, Maurizio Fagnoni, Angelo Albini
ABSTRACT
2-Substituted benzo[b]furans were synthesized by a one-step metal-free photochemical reaction between 2-chlorophenol derivatives and terminal alkynes by tandem formation of an aryl-C and a C-O bond via an aryl cation intermediate. The mild conditions and the application to chlorophenols rather of the more expensive bromo or iodo analogues makes this procedure environmentally convenient.
Sigma-Aldrich
2,3-Benzofuran, 99%