Merck
1,2-dicarbonyl compounds in commonly consumed foods.
Journal of agricultural and food chemistry (2012-06-26)
Julia Degen, Michael Hellwig, Thomas Henle
ABSTRACT
1,2-Dicarbonyl compounds, formed from carbohydrates during thermal processing in the course of caramelization and Maillard reactions, are intensively discussed as precursors for advanced glycation endproducts in foods and in vivo. To obtain information about the uptake of individual compounds with commonly consumed foods, a comprehensive analysis of the content of 3-deoxyglucosone (3-DG), 3-deoxygalactosone (3-DGal), and methylglyoxal (MGO) together with 5-hydroxymethylfurfural (HMF) in 173 food items like bakery products, pasta, nonalcoholic and alcoholic beverages, sweet spreads, and condiments was performed. Following suitable cleanup procedures, 1,2-dicarbonyl compounds were quantitated after derivatization with o-phenylenediamine via RP-HPLC with UV detection. 3-DG proved to be the predominant 1,2-dicarbonyl compound with concentrations up to 410 mg/L in fruit juices, 2622 mg/L in balsamic vinegars, and 385 mg/kg in cookies, thus exceeding the corresponding concentrations of HMF. 3-DGal was found to be of relevance in many foods even in the absence of galactose. MGO was only of minor quantitative importance in all foods studied, except for manuka honey. Dietary intake was estimated to range between 20 and 160 mg/day for 3-DG and 5 and 20 mg/day for MGO, respectively.
Sigma-Aldrich
3-Deoxyglucosone, ≥75% (TLC)