Merck
  • Temperature-sensitive gels for intratumoral delivery of β-lapachone: effect of cyclodextrins and ethanol.
Temperature-sensitive gels for intratumoral delivery of β-lapachone: effect of cyclodextrins and ethanol.
TheScientificWorldJournal (2012-05-26)
Marcilio S S Cunha-Filho, Carmen Alvarez-Lorenzo, Ramón Martínez-Pacheco, Mariana Landin
ABSTRACT
This work evaluated the potential of Pluronics (varieties F127 and P123) in combination with solubilizing agents to be used as syringeable in situ gelling depots of intratumoral β-lapachone (βLAP). Pluronic dispersions prepared at various concentrations in the absence and the presence of ethanol and randomly methylated β-cyclodextrin (RMβCD) were characterized regarding their rheological properties, drug solubilization capacity, and in vitro release. Pluronic F127 (18-23%) formulations combined high ability to solubilize βLAP (enhancement solubility factor up to 50), adequate gel temperature range (over 25 °C), and gel strength at 37 °C enough to guarantee the permanence of the formulation in the administration site for a period of time. βLAP release rate was finely tuned by the concentration of the polymer and the addition of RMβCD (diffusion coefficient ranging between 9 and 69 μg · cm(-2)). The ethanol increases βLAP release rate but simultaneously led to weak gels. This paper shows that βLAP formulations involving temperature-reversible Pluronic gels may be suitable for intratumoral drug delivery purposes.
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,000
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Poloxamer 188, micronized
Sigma-Aldrich
Synperonic® PE/P84
Sigma-Aldrich
Poloxamer 188
Sigma-Aldrich
Poloxamer 407, micronized
Sigma-Aldrich
Poloxamer 407, oxyethylene 71.5-74.9 %
Sigma-Aldrich
Synperonic® F 108, surfactant, non-ionic
Sigma-Aldrich
Poloxamer 407, purified, non-ionic
Sigma-Aldrich
Synperonic® PE P105, surfactant
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,100
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~4,400
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~3,300
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~5,800
Sigma-Aldrich
Poly(ethylene glycol-ran-propylene glycol) monobutyl ether, average Mn ~3,900
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~8,400
Sigma-Aldrich
β-Lapachone, ≥98% (TLC)
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Poloxamer 188, solid
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,900
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,900
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600