Merck
Handling the limit of detection by extrapolation.
Statistics in medicine (2012-04-26)
Nicholas T Longford
ABSTRACT
A general method of estimation with a variable observed subject to a limit of detection is introduced. It is based on extrapolation of the estimates obtained by increasing the limit of detection. Theoretical arguments support the method in some special cases, and it is explored by simulations. Several examples are presented and adaptations for other kinds of censoring explored.
Sigma-Aldrich
Oltipraz, ≥98% (HPLC), powder