Merck
  • Efficiency improvement of organic solar cells by tuning hole transport layer with germanium oxide.
Efficiency improvement of organic solar cells by tuning hole transport layer with germanium oxide.
Journal of nanoscience and nanotechnology (2012-04-25)
Moon Kee Choi, Ju-Hyung Kim, Hyunsik Yoon, Dongha Tahk, Soonmin Seo, Kyusoon Shin, Hong H Lee
ABSTRACT
Improving optical property is critical for optimizing the power conversion efficiency of organic solar cells. In the present research, we show that modification of poly(3,4-ethylenedioxythiophene): poly(styrenesulfonate) (PEDOT:PSS) layer with GeO2 leads to 15% improvement of power conversion efficiency in a polymer solar cells through enhancement of short circuit currents. Modified PEDOT:PSS layer with optimized concentration of GeO2 assists active layer absorbing much light by playing a role of optical spacer. Using AFM and grazing incidence X-ray diffraction (GIXD) data, we also present the evidence that an addition of GeO2 does not affect crystallinity of active layer.
Sigma-Aldrich
Germanium(IV) oxide, (crystalline powder), 99.998% trace metals basis
Sigma-Aldrich
Germanium(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Germanium(IV) oxide, powder, 99.999% trace metals basis