Merck
Evaluation, synthesis and characterization of tacrolimus impurities.
The Journal of antibiotics (2012-04-19)
Patrizia Ferraboschi, Diego Colombo, Maria De Mieri, Paride Grisenti
ABSTRACT
Tacrolimus is an immunosuppressant macrolactam of fermentative origin. By means of HPLC, LC-MS and NMR analyses, coupled with the reference standard synthesis, the main impurities of tacrolimus bulk drug samples were identified and their chemical-physical properties reported. Known ascomycin and tautomers I and II were detected. The correct relative retention time HPLC value of 39,40-dihydro tacrolimus was established. The not described 23,24-anhydro derivative was detected and completely characterized. A full characterization of ascomycin and 39,40-dihydro tacrolimus was also reported.
Sigma-Aldrich
Ascomycin from Streptomyces hygroscopicus var. ascomyceticus
Supelco
Ascomycin solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®