Merck
  • Intramolecular [2+2] cycloaddition of homoallylketenes to bicyclo[2.1.1]hexan-5-ones.
Intramolecular [2+2] cycloaddition of homoallylketenes to bicyclo[2.1.1]hexan-5-ones.
Chemical & pharmaceutical bulletin (2012-04-03)
Takaya Hoshikawa, Kei Tanji, Jun-ichi Matsuo, Hiroyuki Ishibashi
ABSTRACT
Intramolecular [2+2] cycloaddition of γ,δ-unsaturated ketenes derived from hex-5-enoyl chloride derivatives gave bicyclo[2.1.1]hexan-5-ones with complete regioselectivity.
Sigma-Aldrich
2-Hexanone, reagent grade, 98%
Supelco
2-Hexanone, analytical standard
Supelco
2-Hexanone, analytical standard