Merck
  • Monte Carlo calculation of response functions to gamma-ray point sources for a spherical NaI(Tl) detector.
Monte Carlo calculation of response functions to gamma-ray point sources for a spherical NaI(Tl) detector.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2012-03-20)
Chul-Young Yi, Suck-Ho Hah
ABSTRACT
Response functions of a 7.62 cm-diameter spherical NaI(Tl) detector to gamma-ray point sources in the energy range up to 1.5 MeV were calculated by means of the Monte Carlo method using PENELOPE-2006 (Salvat et al., 2006). The detector materials and dimensions were modeled realistically. The calculated response functions agreed well with the experimental spectra.
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium iodide, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis