Merck
  • Inhibitory effect of verbascoside isolated from Buddleja brasiliensis Jacq. ex Spreng on prolyl oligopeptidase activity.
Inhibitory effect of verbascoside isolated from Buddleja brasiliensis Jacq. ex Spreng on prolyl oligopeptidase activity.
Phytotherapy research : PTR (2012-01-26)
Augusto G Filho, Ademir F Morel, Luciana Adolpho, Vinícius Ilha, Ernest Giralt, Teresa Tarragó, Ionara I Dalcol
ABSTRACT
The phenylpropanoid glycoside verbascoside [2-(3,4-dihydroxyphenylethyl)-1-O-α-L-rhamnopyranosyl-(1→3)-β-D-(4-O-caffeyl)-glucopyranoside] (1) has been isolated as the main constituent of the crude extract of Buddleja brasiliensis Jacq. ex Spreng from Southern Brazil. The crude extract, main fractions and the compound 1 were evaluated for inhibition of the enzymes acetylcholinesterase (AChE), dipeptidyl peptidase-IV (DPP-IV) and prolyl oligopeptidase (POP). Compound 1 showed weak activity against DPP-IV with an IC(50) > 150 µM and was inactive against AChE, with a pMIQ determined by bioautography of 9.6. In contrast, 1 displayed significant inhibition of POP in a dose-dependent manner with an IC(50) value of 1.3 ± 0.2 µM, similar to the positive control, baicalin, with a POP IC(50) of 12 ± 3 µM.
Product Description
Verbascoside, primary reference standard
Sigma-Aldrich
Verbascoside, ≥99% (HPLC)