Merck
  • Reactions of a sulfonamide antimicrobial with model humic constituents: assessing pathways and stability of covalent bonding.
Reactions of a sulfonamide antimicrobial with model humic constituents: assessing pathways and stability of covalent bonding.
Environmental science & technology (2012-01-21)
Anna Gulkowska, Martin Krauss, Daniel Rentsch, Juliane Hollender
ABSTRACT
The mechanism of covalent bond formation of the model sulfonamide sulfathiazole (STZ) and the stronger nucleophile para-ethoxyaniline was studied in reactions with model humic acid constituents (quinones and other carbonyl compounds) in the absence and presence of laccase. As revealed by high resolution mass spectrometry, the initial bonding of STZ occurred by 1,2- and 1,4-nucleophilic additions of the aromatic amino group to quinones resulting in imine and anilinoquinone formation, respectively. Experiments using the radical scavenger tert-butyl-alcohol provided the same products and similar formation rates as those without scavenger indicating that probably not radical coupling reactions were responsible for the initial covalent bond formation. No addition with nonquinone carbonyl compounds occurred within 76 days except for a slow 1,4-addition to the β-unsaturated carbonyl 1-penten-3-one. The stability of covalent bonds against desorption and pressurized liquid extraction (PLE) was assessed. The recovery rates showed no systematic differences in STZ extractability between the two product types. This suggests that the strength of bonding is not controlled by the initial type of bond, but by the extent of subsequent incorporation of the reaction product into the formed polymer. This incorporation was monitored for (15)N aniline by (1)H-(15)N HMBC NMR spectroscopy. The initial 1,2- and 1,4-addition bonds were replaced by stronger heterocyclic forms with increasing incubation time. These processes could also hold true for soils, and a slow nonextractable residue formation with time could be related to a slow increase of the amount of covalently bound sulfonamide and the strength of bonding.
Product Description
Supelco
Sulfathiazole
Supelco
Sulfathiazole, VETRANAL®, analytical standard
Sigma-Aldrich
4-Ethoxyaniline, 98%