Merck
The volatile compounds in lamb fat are affected by the time of grazing.
Meat science (2011-10-11)
Valentina Vasta, Valentina Ventura, Giuseppe Luciano, Valeria Andronico, Renato I Pagano, Manuel Scerra, Luisa Biondi, Marcella Avondo, Alessandro Priolo
ABSTRACT
This study was designed to assess whether different grazing managements affect the appearance of organic volatile compounds (VOC) in lamb fat. Forty-two lambs were divided into four groups: 9 lambs were restricted in stall and fed concentrate (S group); 12 lambs grazed from 9 am to 5 pm (8h group); 10 lambs grazed from 9 am to 1 pm (4 hAM group) and 11 lambs grazed from 1 pm to 5 pm (4 hPM group). After 72 days on feed the animals were slaughtered and the perirenal fat subjected to SPME-GC/MS analysis of VOC; the data were analyzed by stepwise discriminant analysis. The 6-methyl-2-heptanone and furanone, 2,5-dimethyl, 4-hydroxy-3(2H) allowed discrimination between the 4 hAM and the 4 hPM animals. These are the first results indicating that the time of the day in which the animals are allowed to graze affects the appearance of VOC in lamb fat.
Sigma-Aldrich
trans,trans-2,6-Nonadienal, ≥95%, stabilized, FG
Sigma-Aldrich
trans-2,cis-6-Nonadienal, 95%
Sigma-Aldrich
trans-2,cis-6-Nonadienal, mixture of isomers, ≥96%, stabilized, FCC, FG