Merck
  • Communication: stabilization of radical anions with weakly bound electron in condensed media: a case study of diacetonyl radical anion.
Communication: stabilization of radical anions with weakly bound electron in condensed media: a case study of diacetonyl radical anion.
The Journal of chemical physics (2011-09-22)
Elizaveta V Saenko, Dimitri N Laikov, Irina A Baranova, Vladimir I Feldman
ABSTRACT
The radical anion resulting from electron capture by diacetonyl molecule has been characterized by EPR and optical absorption spectroscopy in glassy ether matrices at 77 K. In non-polar alkane glasses this species was not observed under the same conditions, which confirms the crucial role of matrix interactions in stabilizing this species. Calculations at the MP2 level show the vertical detachment energy to increase gradually from roughly zero for a bare anion to ∼1 eV for the complex involving six ether molecules.
Sigma-Aldrich
2,5-Hexanedione, ≥98%