Merck
  • Phagostimulatory effect of uptake of PLGA microspheres loaded with rifampicin on alveolar macrophages.
Phagostimulatory effect of uptake of PLGA microspheres loaded with rifampicin on alveolar macrophages.
Colloids and surfaces. B, Biointerfaces (2011-06-28)
Keiji Hirota, Taizo Hasegawa, Takehisa Nakajima, Kimiko Makino, Hiroshi Terada
ABSTRACT
Our previous results on the phagocytic activity of alveolar macrophages (Mϕs) toward poly(lactic-co-glycolic) acid microspheres (PLGA MS) loaded with the anti-tuberculosis agent rifampicin (R-PLGA MS) suggest that the phagocytosis of R-PLGA MS enhances the phagocytic activity of Mϕ cells. To confirm this possibility, we examined the effect of phagocytosis of R-PLGA MS and polystyrene latex (PSL) MS on the phagocytic uptake of fluorescent PSL (F-PSL) MS by cells of the rat alveolar macrophage cell line NR8383 at 37°C. Phagocytic activity was examined in terms of the population of Mϕ cells that had phagocytosed MS (N(total)) and the total number of MS phagocytosed (n(total)) by counting the phagocytic Mϕ cells and the MS ingested in optical microscopic fields. Phagocytosis of R-PLGA MS enhanced about 1.5 times the values of N(total) and n(total) of the phagocytosis of F-PSL MS under the conditions where the phagocytosis of F-PSL MS did not attain the saturated level. In contrast, the phagocytosis of PSL MS did not enhance the phagocytic activity of Mϕ cells toward F-PSL MS. In conclusion, R-PLGA MS are favorable for drug delivery of anti-tuberculosis agents into alveolar Mϕs due to their ability to up-regulate the phagocytosis of MS.
Sigma-Aldrich
Polystyrene, average Mw ~400,000 by GPC
Sigma-Aldrich
Polystyrene, average Mw ~350,000, average Mn ~170,000
Sigma-Aldrich
Polystyrene, average Mw ~192,000
Sigma-Aldrich
Polystyrene, average Mw ~280,000 by GPC