Merck
Oximes as spectrophotometric reagents-a review.
Talanta (1979-06-01)
R B Singh, B S Garg, R P Singh
ABSTRACT
The uses of oximes as spectrophotometric reagents since 1953 are reviewed.
Sigma-Aldrich
Salicylaldoxime, ≥98.0% (NT)
SKU
Pack Size
Availability
Price
Quantity
Supelco
2,3-Butanedione monoxime, for spectrophotometric det. of urea, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethylglyoxime, puriss. p.a., ACS reagent, for the detection of Ni, ≥99.0% (TLC)