Merck
  • Impact of polyphenol metabolites produced by colonic microbiota on expression of COX-2 and GSTT2 in human colon cells (LT97).
Impact of polyphenol metabolites produced by colonic microbiota on expression of COX-2 and GSTT2 in human colon cells (LT97).
Nutrition and cancer (2011-05-21)
Claudia Miene, Anja Weise, Michael Glei
ABSTRACT
Polyphenols may play an important role in colon cancer prevention. After entering the colon, they are subjected to metabolism by the human gut microbiota. The objective of the present study was to analyze the impact of selected intestinal metabolites on modulation of enzymes involved in detoxification and inflammation in human adenoma cells LT97. LT97 cells were incubated with 3,4-dihydroxyphenylacetic acid (ES) and 3-(3,4-dihydroxyphenyl)-propionic acid (PS), metabolites of quercetin and chlorogenic acid/caffeic acid, respectively. The effect on cell number was analyzed using 4'- 6-diamino-2-phenylindole-dihydrochloride (DAPI)-staining. Modulation of glutathione S-transferase T2 (GSTT2) and cyclooxygenase-2 (COX-2) was measured by real-time PCR and Western blot. Comet assay was performed to assess the impact on DNA damage caused by the GSTT2 substrate cumene hydroperoxide (CumOOH). Polyphenol metabolites did not affect cell number but significantly upregulated GSTT2 expression and decreased COX-2. The latter was confirmed via Western blot. CumOOH-induced DNA damage was significantly reduced compared to the control. An upregulation of GSTT2 and downregulation of COX-2 could possibly contribute to the chemopreventive potential of polyphenols after degradation in the gut. Working with polyphenol metabolites is an important prerequisite to better understand the in vivo effects of pure polyphenols.
Sigma-Aldrich
3,4-Dihydroxyhydrocinnamic acid, 98%
Sigma-Aldrich
Cumene hydroperoxide, technical grade, 80%
Sigma-Aldrich
GST T2-2, Recombinant Human