Merck
  • The role of β-adrenergic receptors in the cardioprotective effects of beta-preconditioning (βPC).
The role of β-adrenergic receptors in the cardioprotective effects of beta-preconditioning (βPC).
Cardiovascular drugs and therapy (2011-01-13)
Ruduwaan Salie, Johannes A Moolman, Amanda Lochner
ABSTRACT
To determine the mechanism whereby transient stimulation of the β-adrenergic receptor subtypes (β-AR) elicit cardioprotection against subsequent ischaemia. Isolated rat hearts were subjected to 35 min regional ischaemia (RI) and reperfusion and infarct size (IS) determined. Hearts were preconditioned with 5 min isoproterenol (β1/β2-AR agonist), denopamine (β1-AR agonist), formoterol hemifumarate (β2-AR agonist) or BRL37344 (β3-AR agonist) and 5 min reperfusion. The roles of the β-ARs, NO, PKA, and PI3-K were explored by using selective antagonists/blockers. Pertussis toxin was administered i.p., 48 h prior to experimentation. IS of hearts preconditioned with either isoproterenol, denopamine or formoterol (% of area at risk: 23.6 ± 1.26; 24.52 ± 0.89; 20.74 ± 0.85 respectively) were significantly smaller than that of non-preconditioned hearts (41.7 ± 1.65) and associated with improvement in postischaemic mechanical performance. The β3-AR agonist BRL37344 could not reduce IS. The β1- and β2-AR blockers CGP-20712A and ICI-118551 abolished the reduction in IS and improvement in mechanical recovery during reperfusion induced by isoproterenol preconditioning, while the β3-AR blocker SR59230A was without effect. Both Rp-8-CPT-cAMPs and wortmannin significantly increased IS when administered before and during β1/β2-AR preconditioning and reduced mechanical recovery. PTX pretreatment had no significant effect on the reduction in IS induced by β1/β2-AR or β2-AR preconditioning, but reduced mechanical recovery in β2-AR preconditioning. Similarly the NOS inhibitors L-NAME and LNNA had no effect on IS in β1/β2-AR preconditioning, but depressed mechanical recovery. Protection afforded by β-ARs stimulation, depends on activation of both β1-AR and β2-ARs but not β3-AR. With functional recovery as endpoint, results suggest involvement of NO in β1/β2-AR preconditioning and the Gi protein in β2-AR preconditioning. Both PKA and PI3-K activation were essential for β1/β2-AR cardioprotection.
Sigma-Aldrich
R(−)-Denopamine, ≥98% (HPLC), powder