Merck
  • Spectroscopic characterization and antiproliferative activity on HepG2 human hepatoblastoma cells of flavonoid C-glycosides from Petrorhagia velutina.
Spectroscopic characterization and antiproliferative activity on HepG2 human hepatoblastoma cells of flavonoid C-glycosides from Petrorhagia velutina.
Journal of natural products (2010-11-18)
Severina Pacifico, Monica Scognamiglio, Brigida D'Abrosca, Simona Piccolella, Nikolaos Tsafantakis, Marialuisa Gallicchio, Andreina Ricci, Antonio Fiorentino
ABSTRACT
Eight flavonoid C-glycosides, including three new analogues, have been isolated from leaf and root methanolic extracts of Petrorhagia velutina, a Mediterranean herbaceous plant. The antiproliferative activity against human hepatoblastoma cancer cell line HepG2 has been analyzed by the MTT (3-(4,5-dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium bromide) test. Isoorientin (4) significantly reduces the proliferation of HepG2 cells as determined by the complete conversion of the tetrazolium probe into formazan after 48 h of exposure.
Product Description
Isoorientin, primary reference standard
Sigma-Aldrich
Isoorientin, ≥98% (HPLC)