Merck
  • Novel 6,12-disubstituted chrysene as potent anticancer agent: synthesis, in vitro and in vivo study.
Novel 6,12-disubstituted chrysene as potent anticancer agent: synthesis, in vitro and in vivo study.
European journal of medicinal chemistry (2010-08-13)
Bimal K Banik, Frederick F Becker
ABSTRACT
We describe herein the synthesis of novel 6,12-distributed chrysene as potent anticancer agents. In vitro and in vivo studies are also reported here.
Product Description
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Chrysene, BCR®, certified reference material
Pricing and availability is not currently available.
Supelco
Chrysene, analytical standard
Pricing and availability is not currently available.