Merck
  • Effects of T-type calcium channel blockers on cocaine-induced hyperlocomotion and thalamocortical GABAergic abnormalities in mice.
Effects of T-type calcium channel blockers on cocaine-induced hyperlocomotion and thalamocortical GABAergic abnormalities in mice.
Psychopharmacology (2010-07-24)
Verónica Bisagno, Mariana Raineri, Viviana Peskin, Silvia I Wikinski, Osvaldo D Uchitel, Rodolfo R Llinás, Francisco J Urbano
ABSTRACT
Repetitive cocaine exposure has been shown to induce GABAergic thalamic alterations. Given the key role of T-type (Ca(V)3) calcium channels in thalamocortical physiology, the direct involvement of these calcium channels in cocaine-mediated effects needs to be further explored. The objective of this study was to investigate the effect of T-type calcium channel blockers on acute and repetitive cocaine administration that mediates thalamocortical alterations in mice using three different T-type blockers: 2-octanol, nickel, and mibefradil. During in vitro experiments, whole-cell patch-clamp recordings were conducted in ventrobasal (VB) thalamic neurons from mice treated with acute repetitive cocaine administration (3 x 15 mg/kg, i.p., 1 h apart), under bath application of mibefradil (10 μM), 2-octanol (50 μM), or nickel (200 μM). After systemic administration of T-type calcium channel blockers, we evaluated locomotor activity and also recorded GABAergic neurotransmission onto VB neurons in vitro. Bath-applied mibefradil, 2-octanol, or nickel significantly reduced both GABAergic neurotransmission and T-type currents of VB neurons in cocaine-treated mice. In vivo i.p. pre-administration of either mibefradil (20 mg/kg and 5 mg/kg) or 2-octanol (0.5 mg/kg and 0.07 mg/kg) significantly reduced GABAergic mini frequencies onto VB neurons. Moreover, both mibefradil and 2-octanol were able to decrease cocaine-induced hyperlocomotion. The results shown in this study strongly suggest that T-type calcium channels play a key role in cocaine-mediated GABAergic thalamocortical alterations, and further propose T-type channel blockers as potential targets for future pharmacological strategies aimed at treating cocaine's deleterious effects on physiology and behavior.
Sigma-Aldrich
(±)-2-Octanol, ReagentPlus®, ≥99.5% (GC)
Sigma-Aldrich
(±)-2-Octanol, ≥96.0% (GC)
Sigma-Aldrich
(S)-(+)-2-Octanol, 99%
Sigma-Aldrich
2-Octanol, ≥97%, FG
Sigma-Aldrich
2-Octanol, 97%