Merck
  • Resolution of D- and L-glucoses by chiral N-octyl-beta-D-glycoside-Cu(II) complex adsorbed at the gas/liquid interface of small bubbles.
Resolution of D- and L-glucoses by chiral N-octyl-beta-D-glycoside-Cu(II) complex adsorbed at the gas/liquid interface of small bubbles.
Colloids and surfaces. B, Biointerfaces (2010-07-17)
M Sakai, K Miyazawa, H Jitsumatsu, K Kamio, S Mitsuiki, N Toh, G Sugihara, W Norde
ABSTRACT
A new technique of the jet drop method (JDM) was applied to a chiral molecular discrimination of optically active D- or L-glucose (guest) by chiral N-octyl-beta-D-glycoside (ObetaDG)-Cu(II) complex (host) at the gas/liquid interface of small bubbles. The discrimination of glucoses as the guests is possible using ObetaDG adsorbed at the gas/liquid interface of bubbles where it acts as the host, either in the presence or the absence of Cu(II) ions. In order to make clear the host-guest interaction at the gas/liquid interface, the composition of 5000 top jet drops periodically collected onto a slide glass receiver was analyzed. The relative concentration (eta(i)-1) and the surface excess amount, Gamma(i)(0) of species i such as D- and L-glucoses, and ObetaDG were determined as a function of bubble size, d(b) and bulk concentration C(b). The partition coefficient, Pi(i)=Gamma(i)(0)/C(b) was also evaluated for each component. The adsorption of these materials either in the presence or absence of Cu(II) ions, was assigned to the Freundlich type, and the discrimination of D- and L-glucoses with ObetaDG was evaluated in terms of the Freundlich constant, k(i) and 1/n. The discrimination ability of ObetaDG was also evaluated by determining the equilibrium constants, K(c) of complex formations for the respective glucoses in the presence and absence of Cu(II) ions. It was found that L-glucose can form a more stable complex with ObetaDG-Cu(II).
Sigma-Aldrich
Octyl β-D-glucopyranoside, BioXtra, ≥98% (GC)
Sigma-Aldrich
Octyl β-D-glucopyranoside solution, ≥95% (HPLC), 50 % (w/v) in H2O
Sigma-Aldrich
Octyl β-D-glucopyranoside, ≥98% (GC)
Sigma-Aldrich
Octyl-β-D-glucopyranoside 100 mM solution