Merck
  • Palladium(II)/cationic 2,2'-bipyridyl system as a highly efficient and reusable catalyst for the Mizoroki-Heck reaction in water.
Palladium(II)/cationic 2,2'-bipyridyl system as a highly efficient and reusable catalyst for the Mizoroki-Heck reaction in water.
Molecules (Basel, Switzerland) (2010-01-30)
Shao-Hsien Huang, Jun-Rong Chen, Fu-Yu Tsai
ABSTRACT
A water-soluble and air-stable Pd(NH3)2Cl2/cationic 2,2'-bipyridyl system was found to be a highly-efficient and reusable catalyst for the coupling of aryl iodides and alkenes in neat water using Bu(3)N as a base. The reaction was conducted at 140 degrees C in a sealed tube in air with a catalyst loading as low as 0.0001 mol % for the coupling of activated aryl iodides with butyl and ethyl acrylates, providing the corresponding products in good to excellent yields with very high turnover numbers. In the case of styrene, Mizoroki-Heck coupling products were obtained in good to high yields by using a greater catalyst loading (1 mol %) and TBAB as a phase-transfer agent. After extraction, the residual aqueous solution could be reused several times with only a slight decrease in its activity, making the Mizoroki-Heck reaction "greener".
Sigma-Aldrich
Ethyl acrylate, contains 10-20 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Iodobenzene, 98%
Supelco
Ethyl acrylate, analytical standard
Supelco
Butyl acrylate, analytical standard
Sigma-Aldrich
Butyl acrylate, ≥99%, contains 10-60 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Ethyl acrylate, ≥99.5%, stabilized