Merck
  • Using promoter replacement and selection for loss of heterozygosity to generate an industrially applicable sake yeast strain that homozygously overproduces isoamyl acetate.
Using promoter replacement and selection for loss of heterozygosity to generate an industrially applicable sake yeast strain that homozygously overproduces isoamyl acetate.
Journal of bioscience and bioengineering (2009-10-07)
Hiroshi Sahara, Atsushi Kotaka, Akihiko Kondo, Mitsuyoshi Ueda, Yoji Hata
ABSTRACT
By application of the high-efficiency loss of heterozygosity (HELOH) method for disrupting genes in diploid sake yeast (Kotaka et al., Appl. Microbiol. Biotechnol., 82, 387-395 (2009)), we constructed, from a heterozygous integrant, a homozygous diploid that overexpresses the alcohol acetyltransferase gene ATF2 from the SED1 promoter, without the need for sporulation and mating. Under the conditions of sake brewing, the homozygous integrant produced 1.4 times more isoamyl acetate than the parental, heterozygous strain. Furthermore, the homozygous integrant was more genetically stable than the heterozygous recombinant. Thus, the HELOH method can produce homozygous, recombinant sake yeast that is ready to be grown on an industrial scale using the well-established procedures of sake brewing. The HELOH method, therefore, facilitates genetic modification of this rarely sporulating diploid yeast strain while maintaining those characteristics required for industrial applications.
Sigma-Aldrich
Isopentyl acetate, anhydrous, ≥99%
Sigma-Aldrich
Isopentyl acetate, mixture of isomers, reagent grade, 98%
Supelco
Isopentyl acetate, analytical standard
Sigma-Aldrich
Isoamyl acetate, ≥95%, FCC, FG
Sigma-Aldrich
Isoamyl acetate, natural, ≥97%, FCC, FG