Merck
  • Interactive effect of organic amendment and environmental factors on degradation of 1,3-dichloropropene and chloropicrin in soil.
Interactive effect of organic amendment and environmental factors on degradation of 1,3-dichloropropene and chloropicrin in soil.
Journal of agricultural and food chemistry (2009-09-03)
Ruijun Qin, Suduan Gao, Husein Ajwa, Bradley D Hanson, Thomas J Trout, Dong Wang, Mingxin Guo
ABSTRACT
Soil organic matter is an important factor affecting the fate of soil fumigants; therefore, the addition of organic amendments to surface soils could reduce fumigant emissions by accelerating fumigant degradation. Experiments were conducted to determine the degradation of fumigants [a mixture of cis- and trans-1,3-dichloropropene (1,3-D) and chloropicrin (CP), a similar composition as in Telone C35] in soils with organic amendment under a range of soil moisture, temperature, sterilization, and texture conditions. Degradation of the fumigants followed availability-adjusted first-order or pseudo-first-order kinetics with slower degradation of 1,3-D than CP. Increasing soil water content from 5 to 17.5% (w/w) slightly increased the degradation of 1,3-D, but not that of CP. Five different organic amendments at 5% (w/w) increased fumigant degradation 1.4-6.3-fold in this study. The degradation of both fumigants was accelerated with increasing amount of organic material (OM). Little interaction between soil moisture and OM was observed. Autoclave sterilization of soils did not reduce degradation of either fumigant; however, increasing the incubation temperature from 10 to 45 degrees C accelerated fumigant degradation 5-14 times. Soil texture did not affect 1,3-D degradation, but CP degraded more rapidly in finer-textured soil. These results suggest that OM type and rate and soil temperature are the most important factors affecting the degradation of 1,3-D and CP.
Product Description
Supelco
trans-1,3-Dichloropropene, analytical standard
Sigma-Aldrich
cis-1,3-Dichloropropene, 97%
Sigma-Aldrich
1,3-Dichloropropene, 90%, technical grade