Merck
  • Palladium-catalyzed decarboxylative rearrangements of allyl 2,2,2-trifluoroethyl malonates: direct access to homoallylic esters.
Palladium-catalyzed decarboxylative rearrangements of allyl 2,2,2-trifluoroethyl malonates: direct access to homoallylic esters.
Organic letters (2009-08-22)
Lawrence P Tardibono, Jerod Patzner, Cara Cesario, Marvin J Miller
ABSTRACT
Homoallylic esters are obtained in a single transformation from allyl 2,2,2-trifluoroethyl malonates by using a Pd(0) catalyst. Facile decarboxylation of allyl 2,2,2-trifluoroethyl malonates is attributed to a decrease in pK(a) compared to allyl methyl malonates. Subsequent reduction of the homoallylic 2,2,2-trifluoroethyl ester provides a (hydroxyethyl)cyclopentenyl derivative that represents a key intermediate in the synthesis of carbocyclic nucleosides. A select allyl 2,2,2-trifluoroethyl malonate undergoes a decarboxylative Claisen rearrangement to provide a regioisomeric homoallylic ester.
Sigma-Aldrich
Dimethyl malonate, purum, ≥96.0% (GC)
Sigma-Aldrich
Dimethyl malonate, 98%