Merck
  • Controlled release of dexamethasone acetate from biodegradable and biocompatible polyurethane and polyurethane nanocomposite.
Controlled release of dexamethasone acetate from biodegradable and biocompatible polyurethane and polyurethane nanocomposite.
Journal of drug targeting (2009-06-27)
Gisele Rodrigues Da Silva, Eliane Ayres, Rodrigo Lambert Orefice, Sandra Aparecida L Moura, Denise Carmona Cara, Armando Da Silva Cunha
ABSTRACT
Polyurethanes and polyurethane nanocomposites can be applied to control the release of drugs previously incorporated into these materials. In this study, dexamethasone acetate (ACT) was incorporated into biodegradable and biocompatible polyurethane and polyurethane containing montmorillonite nanoparticles. Fourier transform infrared spectroscopic technique showed no strong interactions between drug and polymers. Data obtained from X-ray diffraction and small angle X-ray scattering indicated that the incorporation of ACT did not disturb the polymer morphology, but montmorillonite led to a less defined phase separation between hard and soft segments of polyurethane. The in vitro release studies demonstrated that nanoparticles increased the rate of ACT release possibly because these particles have a hydrophilic surface that increases the absorption of water and accelerates the hydrolysis of the polymer. The in vivo short-term biocompatibility studies demonstrated adequate interfacial interaction between polyurethane and subcutaneous tissue and a discreet inflammatory response which was completely resolved in 14 days.
Sigma-Aldrich
Dexamethasone 21-acetate, ≥99%
Dexamethasone acetate, European Pharmacopoeia (EP) Reference Standard