Merck
  • Chemical infiltration during atomic layer deposition: metalation of porphyrins as model substrates.
Chemical infiltration during atomic layer deposition: metalation of porphyrins as model substrates.
Angewandte Chemie (International ed. in English) (2009-06-06)
Lianbing Zhang, Avinash J Patil, Le Li, Angelika Schierhorn, Stephen Mann, Ulrich Gösele, Mato Knez
ABSTRACT
New uses for ALD: By applying standard metal oxide atomic layer deposition (ALD) to two types of porphyrins, site-specific chemical infiltration of substrate molecules is achieved: Diethylzinc can diffuse into the interior of porphyrin supramolecular structures and induce metalation of the porphyrin molecules from the vapor phase. A = Ph, p-HO(3)SC(6)H(4).
Sigma-Aldrich
Diethylzinc, ≥52 wt. % Zn basis
Sigma-Aldrich
Diethylzinc solution, 1.0 M in heptane
Sigma-Aldrich
Diethylzinc solution, 15 wt. % in toluene
Sigma-Aldrich
Diethylzinc solution, 1.0 M in hexanes