Merck
  • Methyl mercapturate synthesis: an efficient, convenient and simple method.
Methyl mercapturate synthesis: an efficient, convenient and simple method.
Molecules (Basel, Switzerland) (2008-10-03)
Benoît Cossec, Frédéric Cosnier, Manuella Burgart
ABSTRACT
A safe and simple method for methyl S-arylmercapturate synthesis is described. Thirteen such compounds, to be used afterwards in metabolism studies, have been obtained with yields ranging from 71 to 99.6%. These compounds were obtained using a sulfa-Michael addition and synthesized by adding the corresponding thiophenols to a mixture composed of methyl 2-acetamidoacrylate (MAA), potassium carbonate and a phase transfer catalyst, Aliquat 336. MAA, the initial synthon, was itself isolated in quasi quantitative yield following a fully described synthesis.
Sigma-Aldrich
Methyl 2-acetamidoacrylate, 98%
Pricing and availability is not currently available.