Merck
  • Optical resolution of (+/-)-1,2-Bis(2-methylphenyl)ethylene-1,2-diamine as a chiral framework for 2-iminoimidazolidine with 2-methylphenyl pendant and the guanidine-catalyzed asymmetric michael reaction of tert-butyl diphenyliminoacetate and ethyl acrylate.
Optical resolution of (+/-)-1,2-Bis(2-methylphenyl)ethylene-1,2-diamine as a chiral framework for 2-iminoimidazolidine with 2-methylphenyl pendant and the guanidine-catalyzed asymmetric michael reaction of tert-butyl diphenyliminoacetate and ethyl acrylate.
The Journal of organic chemistry (2007-12-08)
Akemi Ryoda, Nana Yajima, Toyokazu Haga, Takuya Kumamoto, Waka Nakanishi, Masatoshi Kawahata, Kentaro Yamaguchi, Tsutomu Ishikawa
ABSTRACT
(+/-)-1,2-Bis(2-methylphenyl)ethylene-1,2-diamine, prepared from benzil and ammonium acetate, was optically resolved as a chiral framework for 2-(1-benzyl-2-hydroxyethyl)imino-1,3-dimethylimidazolidine with 2-methylphenyl pendants at the 4,5-positions. Catalysis ability of the 1,3-dimethyl-4,5-bis(2-methylphenyl)imidazolidine and the related 1,3-dibenzyl-4,5-diphenylimidazolidine was examined in the asymmetric Michael reaction of t-butyl diphenyliminoacetate and ethyl acrylate.
Sigma-Aldrich
Ethyl acrylate, contains 10-20 ppm MEHQ as inhibitor, 99%
Supelco
Ethyl acrylate, analytical standard
Sigma-Aldrich
Ethyl acrylate, ≥99.5%, stabilized