Merck
Fatty acid composition of Abies pindrow (West Himalayan fir).
Pakistan journal of pharmaceutical sciences (2007-03-06)
Dadu Khan Burdi, M Qasim Samejo, M Iqbal Bhanger, Khalid M Khan
ABSTRACT
The leaves of Abies pindrow, collected from Murree Hills, Punjab (Pakistan) revealed the presence of eleven fatty acids including eight saturated and three unsaturated fatty acids. They ranged from C(14) to C(24) and were detected as methyl esters by GC-MS technique. The saturated fatty acids were present in much greater proportion than unsaturated ones. Isopalmatic acid was found to be major saturated fatty acid and the oleic acid as predominant unsaturated acid. (+)-14-Methyl palmatic acid and (+)-Isosteric acid were the next higher saturated and unsaturated fatty acids respectively.
Sigma-Aldrich
Methyl palmitate, ≥99% (capillary GC)
Sigma-Aldrich
Methyl palmitate, ≥97%
Supelco
Methyl palmitate, analytical standard