Merck
  • Unanticipated differences between alpha- and gamma-diaminobutyric acid-linked hairpin polyamide-alkylator conjugates.
Unanticipated differences between alpha- and gamma-diaminobutyric acid-linked hairpin polyamide-alkylator conjugates.
Nucleic acids research (2006-12-16)
Sherry M Tsai, Michelle E Farkas, C James Chou, Joel M Gottesfeld, Peter B Dervan
ABSTRACT
Hairpin polyamide-chlorambucil conjugates containing an alpha-diaminobutyric acid (alpha-DABA) turn moiety are compared to their constitutional isomers containing the well-characterized gamma-DABA turn. Although the DNA-binding properties of unconjugated polyamides are similar, the alpha-DABA conjugates display increased alkylation specificity and decreased rate of reaction. Treatment of a human colon carcinoma cell line with alpha-DABA versus gamma-DABA hairpin conjugates shows only slight differences in toxicities while producing similar effects on cell morphology and G2/M stage cell cycle arrest. However, striking differences in animal toxicity between the two classes are observed. Although mice treated with an alpha-DABA hairpin polyamide do not differ significantly from control mice, the analogous gamma-DABA hairpin is lethal. This dramatic difference from a subtle structural change would not have been predicted.
Sigma-Aldrich
L-2,4-Diaminobutyric acid dihydrochloride, ≥95.0%