Merck
  • Intracoronary ghrelin infusion decreases coronary blood flow in anesthetized pigs.
Intracoronary ghrelin infusion decreases coronary blood flow in anesthetized pigs.
Endocrinology (2006-11-18)
Elena Grossini, Claudio Molinari, David A S G Mary, Ezio Ghigo, Gianni Bona, Giovanni Vacca
ABSTRACT
The peptide ghrelin has been linked to the atherosclerotic process and coronary artery disease. We planned to study, for the first time, the primary effects of ghrelin on the intact coronary circulation and determine the mechanisms involved. In 24 sodium pentobarbitone-anesthetized pigs, changes in anterior descending coronary blood flow caused by intracoronary infusion of ghrelin at constant heart rate and arterial pressure were assessed using electromagnetic flowmeters. In 20 pigs, intracoronary infusion of ghrelin decreased coronary blood flow without affecting left ventricular maximum rate of change of left ventricular systolic pressure (dP/dt(max)), filling pressures of the heart or plasma levels of GH. In four pigs, this decrease was graded by step increments of infused dose of the hormone. The mechanisms of the above response were studied in the 20 pigs by repeating the experiment after coronary flow had returned to the control values observed before infusion. The ghrelin-induced coronary vasoconstriction was not affected by iv atropine (five pigs) or phentolamine (five pigs). This response was abolished by iv butoxamine (five pigs) and intracoronary N(omega)-nitro-l-arginine methyl ester (five pigs), even after reversing the increase in arterial pressure and coronary vascular resistance caused by the two blocking agents with iv infusion of papaverine. The present study showed that intracoronary infusion of ghrelin primarily caused coronary vasoconstriction. The mechanisms of this response were shown to involve the inhibition of a vasodilatory beta(2)-adrenergic receptor-mediated effect related to the release of nitric oxide.
Product Description
Supelco
Butoxamine hydrochloride, analytical standard, for drug analysis, mixture of diastereomers
Pricing and availability is not currently available.